Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt77
Entrez Gene ID
406226
Gene Name
keratin 77, type II
Gene Aliases
Kb39
Chromosome Location
Chr.7: 143461423 - 143473840 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008807.1 NP_001008807.1 1 - 2 555 154
XM_006242435.2 XP_006242497.1 1 - 2 698 154

View Full Product Details