Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-CE11
Entrez Gene ID
414791
Gene Name
RT1 class I, locus CE11
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008834.1 NP_001008834.1 6 - 7 1056 94

View Full Product Details