The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013163.2 | NP_001013181.2 | 1 - 1 | 255 | 75 |
GenBank mRNA | DQ191782.1 | - | 1 - 1 | 251 | 75 |
DQ191783.1 | - | 1 - 1 | 238 | 75 | |
DQ191784.1 | - | 1 - 1 | 255 | 75 | |
DQ191785.1 | - | 1 - 1 | 235 | 75 | |
DQ191786.1 | - | 1 - 1 | 255 | 75 |