Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem80
Entrez Gene ID
309109
Gene Name
transmembrane protein 80
Gene Aliases
RGD1308399
Chromosome Location
Chr.1: 214317300 - 214326293 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045203.1 3 - 4 877 101
GenBank mRNA BC083751.1 - 3 - 4 495 101

View Full Product Details