The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024324.1 | NP_001019495.1 | 2 - 3 | 479 | 62 |
XM_006236223.2 | XP_006236285.1 | 4 - 5 | 841 | 62 | |
XM_006236224.2 | XP_006236286.1 | 4 - 5 | 841 | 62 | |
XM_006236225.2 | XP_006236287.1 | 4 - 5 | 841 | 62 | |
XM_008762818.1 | XP_008761040.1 | 4 - 5 | 841 | 62 | |
GenBank mRNA | BC079297.1 | - | 2 - 3 | 479 | 62 |