Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tpra1
Entrez Gene ID
85494
Gene Name
transmembrane protein, adipocyte asscociated 1
Gene Aliases
Gpr175, Tpra40
Chromosome Location
Chr.4: 120843376 - 120854534 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF292116.1 - 10 - 10 934 75

View Full Product Details