Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Kcna1
Entrez Gene ID
24520
Gene Name
potassium voltage-gated channel, shaker-related subfamily, member 1
Gene Aliases
Kcna, Kcpvd, Kv1.1
Chromosome Location
Chr.4: 159190781 - 159192526 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA X12589.1 - - 3044 116

View Full Product Details