The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022857.1 | NP_074048.1 | 2 - 2 | 2047 | 142 |
GenBank mRNA | L31882.1 | - | 2 - 2 | 2047 | 142 |