Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhox11
Entrez Gene ID
298346
Gene Name
reproductive homeobox 11
Gene Aliases
-
Chromosome Location
Chr.X: 124228270 - 124236517 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024873.1 NP_001020044.1 2 - 3 421 84
GenBank mRNA DQ058658.1 - 2 - 3 421 84

View Full Product Details