Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dhrs4
Entrez Gene ID
266686
Gene Name
dehydrogenase/reductase (SDR family) member 4
Gene Aliases
PSCD
Chromosome Location
Chr.15: 34155255 - 34167073 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153315.2 NP_695227.2 5 - 6 662 77
GenBank mRNA AB062758.1 - 5 - 6 478 77
BC070961.1 - 5 - 6 662 77

View Full Product Details