Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chst15
Entrez Gene ID
286974
Gene Name
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Gene Aliases
GalNAc4S6ST, Galnac4s-6st
Chromosome Location
Chr.1: 204243396 - 204328970 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230430.2 XP_006230492.1 2 - 3 2122 85
XM_006230431.2 XP_006230493.1 2 - 3 2862 85
XM_006230433.2 XP_006230495.1 2 - 3 2357 85
XM_008759868.1 XP_008758090.1 2 - 3 1118 85
XM_008759869.1 XP_008758091.1 2 - 3 2861 85
GenBank mRNA BC085687.1 - 1 - 2 816 85

View Full Product Details