The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013093.1 | NP_001013111.1 | 2 - 2 | 775 | 66 |
GenBank mRNA | BC079223.1 | - | 2 - 2 | 775 | 66 |