The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013886.2 | NP_001013908.2 | 4 - 4 | 834 | 104 |
GenBank mRNA | BC105754.1 | - | 4 - 4 | 834 | 104 |
X03369.1 | - | 4 - 4 | 743 | 104 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109119.1 | NP_001102589.1 | - | 812 | 104 |
XM_008771582.1 | XP_008769804.1 | - | 837 | 104 |