The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017375.2 | NP_001017375.2 | 9 - 10 | 2054 | 63 |
GenBank mRNA | BC099116.1 | - | 9 - 10 | 1997 | 63 |