The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133425.3 | NP_596916.1 | 3 - 4 | 1026 | 85 |
GenBank mRNA | AF407168.3 | - | 3 - 4 | 1026 | 85 |
BC086978.1 | - | 3 - 4 | 499 | 85 |