The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022300.1 | NP_071636.1 | 2 - 2 | 743 | 90 |
GenBank mRNA | D14441.1 | - | 2 - 2 | 743 | 90 |