The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153726.1 | NP_714948.1 | 9 - 9 | 1065 | 152 |
GenBank mRNA | AY102036.1 | - | 9 - 9 | 1065 | 152 |
AY747628.1 | - | 10 - 10 | 1116 | 152 |