Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpr82
Entrez Gene ID
100362441
Gene Name
G protein-coupled receptor 82
Gene Aliases
-
Chromosome Location
Chr.X: 9997816 - 10001036 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773064.1 XP_008771286.1 1 - 1 2070 113

View Full Product Details