The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100943.1 | NP_001094413.1 | 1 - 1 | 453 | 124 |
NM_001114392.1 | NP_001107864.1 | 1 - 1 | 653 | 124 |