Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cyp3a23/3a1
- Entrez Gene ID
-
25642
- Gene Name
- cytochrome P450, family 3, subfamily a, polypeptide 23/polypeptide 1
- Gene Aliases
-
CYP, CYP3A23, Cyp3a1, Cyp3a3, RL33, cDEX
- Chromosome Location
-
Chr.12: 11053888 - 11082742 on Build Rnor_6.0
- Gene Symbol
- LOC100910877
- Entrez Gene ID
-
100910877
- Gene Name
- cytochrome P450 3A1-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 11703602 - 11733127 on Build Rnor_6.0
View Full Product Details