Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ceacam6
- Entrez Gene ID
-
100125369
- Gene Name
- carcinoembryonic antigen-related cell adhesion molecule 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 81683500 - 81701230 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_045097.1 |
|
9 - 10 |
1689 |
67 |
GenBank mRNA |
BC078962.1 |
- |
9 - 10 |
1689 |
67 |
- Gene Symbol
- LOC103689938
- Entrez Gene ID
-
103689938
- Gene Name
- carcinoembryonic antigen-related cell adhesion molecule 1-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 83043478 - 83061421 on Build Rnor_6.0
View Full Product Details