The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107639.2 | NP_001101109.2 | 3 - 4 | 630 | 65 |
XM_008772991.1 | XP_008771213.1 | 4 - 5 | 595 | 65 |