Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r49
Entrez Gene ID
688392
Gene Name
vomeronasal 2 receptor, 49
Gene Aliases
-
Chromosome Location
Chr.6: 56732891 - 56775529 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099515.1 NP_001092985.1 1 - 2 212 100

View Full Product Details