Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkr-p1c
Entrez Gene ID
678513
Gene Name
killer cell lectin-like receptor subfamily B member
Gene Aliases
Klrb1b, NKR-P1B, NKRP1-B, Nkrp1b
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040189.2 NP_001035279.1 6 - 6 1247 131
GenBank mRNA DQ157010.1 - 6 - 6 1247 131

View Full Product Details