Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom2r19
Entrez Gene ID
690729
Gene Name
vomeronasal 2 receptor, 19
Gene Aliases
-
Chromosome Location
Chr.1: 65409829 - 65422705 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099492.1 NP_001092962.1 6 - 6 2147 154

Gene Symbol
LOC100910798
Entrez Gene ID
100910798
Gene Name
vomeronasal type-2 receptor 26-like
Gene Aliases
-
Chromosome Location
Chr.8: 4405227 - 4419385 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750426.3 XP_003750474.2 - 2147 154

Gene Symbol
Vom2r21
Entrez Gene ID
100101439
Gene Name
vomeronasal 2 receptor, 21
Gene Aliases
-
Chromosome Location
Chr.1: 65509270 - 65512227 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006228107.2 XP_006228169.2 2 - 2 798 154
GenBank mRNA AF053991.1 - 1 - 1 184 154

View Full Product Details