Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom2r78
Entrez Gene ID
367480
Gene Name
vomeronasal 2 receptor, 78
Gene Aliases
-
Chromosome Location
Chr.1: 67503956 - 67522575 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099659.1 NP_001093129.1 - 350 140

Gene Symbol
Vom2r32
Entrez Gene ID
286981
Gene Name
vomeronasal 2 receptor, 32
Gene Aliases
-
Chromosome Location
Chr.1: 67819055 - 67853582 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008758879.1 XP_008757101.1 3 - 3 974 140

View Full Product Details