Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC287004
- Entrez Gene ID
-
287004
- Gene Name
- Mg1 protein
- Gene Aliases
-
GB.7, Pinc
- Chromosome Location
-
Chr.9: 80680445 - 80697356 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110640.1 |
|
4 - 5 |
1029 |
145 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DQ099683.1 |
- |
4 - 5 |
994 |
|
Chr. 9 - 72706244 - 72723120 [-] on Build Rnor_6.0 |
145 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
protein unc-80 homolog |
LOC103693241 |
|
-40.6kb
|
Sense |
Chr. - 72663790 - 72665685 on Build Rnor_6.0 |
Downstream |
tensin 1 |
Tns1 |
Tns |
509.3kb
|
Sense |
Chr. - 81191792 - 81401027 on Build Rnor_6.0 |
View Full Product Details