Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctf2
Entrez Gene ID
293515
Gene Name
cardiotrophin 2
Gene Aliases
Ctf2p
Chromosome Location
Chr.1: 199169022 - 199177365 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135800.1 NP_001129272.1 2 - 3 156 74

View Full Product Details