Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gas5
- Entrez Gene ID
-
81714
- Gene Name
- growth arrest specific 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 78852523 - 78855844 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002704.1 |
|
6 - 7 |
189 |
108 |
GenBank mRNA |
FQ233900.1 |
- |
6 - 7 |
216 |
108 |
U77829.1 |
- |
6 - 7 |
189 |
108 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG373751.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
BF414925.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
CO558090.1 |
- |
|
461 |
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
BQ781518.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
FM034982.1 |
- |
|
186 |
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
CN542792.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
BM385132.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
BF287008.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
BG379376.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
CR463429.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
BI303289.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
AI454321.1 |
- |
|
|
|
Chr. - - [-] on Build |
108 |
GenBankmRNA |
CO395348.1 |
- |
|
78 |
|
Chr. - - [-] on Build |
108 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger and BTB domain containing 37 |
Zbtb37 |
RGD1308731 |
-2.0kb
|
Anti-sense |
Chr. - 78836491 - 78852211 on Build Rnor_6.0 |
Downstream |
aspartyl-tRNA synthetase 2 (mitochondrial) |
Dars2 |
RGD1308286 |
3.0kb
|
Anti-sense |
Chr. - 78857638 - 78885464 on Build Rnor_6.0 |
View Full Product Details