Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rmrp
- Entrez Gene ID
-
29536
- Gene Name
- RNA component of mitochondrial RNA processing endoribonuclease
- Gene Aliases
-
Rmrp1
- Chromosome Location
-
Chr.15: 27738989 - 27739245 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002703.1 |
|
1 - 1 |
144 |
104 |
GenBank mRNA |
L08800.1 |
- |
1 - 1 |
172 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cyclin B1 interacting protein 1, E3 ubiquitin protein ligase |
Ccnb1ip1 |
|
-2.7kb
|
Sense |
Chr. - 27720220 - 27736308 on Build Rnor_6.0 |
Downstream |
poly (ADP-ribose) polymerase 2 |
Parp2 |
Adprtl2 |
171.0
|
Anti-sense |
Chr. - 27739416 - 27749650 on Build Rnor_6.0 |
View Full Product Details