The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100728.1 | NP_001094198.1 | 4 - 5 | 507 | 64 |
GenBank mRNA | BC091255.1 | - | 4 - 5 | 481 | 64 |