The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136124.1 | NP_001129596.1 | 2 - 2 | 446 | 113 |
GenBank mRNA | AF164039.1 | - | 1 - 1 | 937 | 113 |
BC168774.1 | - | 2 - 2 | 446 | 113 |