Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lst1
Entrez Gene ID
64569
Gene Name
leukocyte specific transcript 1
Gene Aliases
B144
Chromosome Location
Chr.20: 5175291 - 5179353 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022634.2 NP_072156.2 2 - 3 89 67
XM_006256078.2 XP_006256140.1 2 - 3 230 67
XM_006256079.2 XP_006256141.1 2 - 3 230 67

View Full Product Details