Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pim2
Entrez Gene ID
317366
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
Gene Aliases
-
Chromosome Location
Chr.X: 15462621 - 15467890 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172103.1 NP_001165574.1 5 - 6 957 69

View Full Product Details