Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tlx2
Entrez Gene ID
680117
Gene Name
T-cell leukemia homeobox 2
Gene Aliases
Hox11L.1
Chromosome Location
Chr.4: 114765221 - 114768243 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172125.1 NP_001165596.1 1 - 2 618 88

Gene Symbol
NEWGENE_1595506
Entrez Gene ID
103692164
Gene Name
T-cell leukemia homeobox 2
Gene Aliases
Tlx2
Chromosome Location
Chr.4: 113899376 - 113902237 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763031.1 XP_008761253.1 1 - 2 456 88

View Full Product Details