Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl26
Entrez Gene ID
290657
Gene Name
kelch-like family member 26
Gene Aliases
RGD1312034
Chromosome Location
Chr.16: 20704807 - 20729882 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106075.1 NP_001099545.1 2 - 3 310 82

View Full Product Details