Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb42
Entrez Gene ID
691556
Gene Name
zinc finger and BTB domain containing 42
Gene Aliases
-
Chromosome Location
Chr.6: 137238893 - 137247115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126302.1 NP_001119774.1 1 - 2 99 82
GenBank mRNA BC161864.1 - 1 - 2 99 82

View Full Product Details