The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135020.1 | NP_001128492.1 | 3 - 4 | 274 | 89 |
GenBank mRNA | BC168720.1 | - | - | 223 | 89 |
BC168779.1 | - | 3 - 4 | 244 | 89 | |
CB313385.1 | - | 3 - 4 | 275 | 89 | |
CR464634.1 | - | 1 - 2 | 89 | ||
DV719731.1 | - | 3 - 4 | 274 | 89 | |
FQ221821.1 | - | 3 - 4 | 280 | 89 |