Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc6a8
Entrez Gene ID
50690
Gene Name
solute carrier family 6 (neurotransmitter transporter), member 8
Gene Aliases
CHOT1, CHT1, CRT, CT1
Chromosome Location
Chr.X: 157129987 - 157139322 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017348.2 NP_059044.1 11 - 12 2179 96
XM_006229611.1 XP_006229673.1 10 - 11 2076 96
GenBank mRNA BC168238.1 - 11 - 12 2157 96

View Full Product Details