The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024138.1 | NP_077052.1 | 2 - 3 | 325 | 100 |
GenBank mRNA | DQ120497.1 | - | 1 - 2 | 83 | 100 |
DQ120498.1 | - | 1 - 2 | 83 | 100 | |
FQ211653.1 | - | - | 305 | 100 | |
L23219.1 | - | 2 - 3 | 325 | 100 |