The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017243.2 | NP_058939.1 | 7 - 7 | 1346 | 105 |
GenBank mRNA | CO405720.1 | - | 2 - 2 | 496 | 105 |
X16554.1 | - | 7 - 7 | 1343 | 105 |