The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012360.1 | NP_001012360.1 | 3 - 3 | 1144 | 59 |
XM_006253236.2 | XP_006253298.1 | 4 - 4 | 1529 | 59 | |
GenBank mRNA | AJ879584.1 | - | 3 - 3 | 1144 | 59 |
AJ880010.1 | - | 3 - 3 | 1144 | 59 |