Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Slc2a7
Entrez Gene ID
100362644
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 7
Gene Aliases
-
Chromosome Location
Chr.5: 167188086 - 167211608 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101020.1 NP_001094490.1 - 956 68
GenBank mRNA EF501821.1 - - 956 68

View Full Product Details