Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir19b1
- Entrez Gene ID
-
100314186
- Gene Name
- microRNA 19b-1
- Gene Aliases
-
Mir19b-1, rno-mir-19b-1
- Chromosome Location
-
Chr.15: 100180464 - 100180550 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_031820.1 |
|
1 - 1 |
42 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 20a |
Mir20a |
rno-mir-20a |
-46.0
|
Sense |
Chr. - 100180334 - 100180418 on Build Rnor_6.0 |
Downstream |
microRNA 92a-1 |
Mir92a1 |
Mir92a-1, rno-mir-92a-1 |
36.0
|
Sense |
Chr. - 100180586 - 100180663 on Build Rnor_6.0 |
View Full Product Details