Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir25
- Entrez Gene ID
-
100314004
- Gene Name
- microRNA mir-25
- Gene Aliases
-
rno-mir-25
- Chromosome Location
-
Chr.12: 19307340 - 19307423 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_031829.1 |
|
1 - 1 |
41 |
55 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
COP9 signalosome subunit 6 |
Cops6 |
|
-1.1kb
|
Anti-sense |
Chr. - 19303387 - 19306226 on Build Rnor_6.0 |
Downstream |
microRNA mir-93 |
Mir93 |
rno-mir-93 |
120.0
|
Sense |
Chr. - 19307543 - 19307629 on Build Rnor_6.0 |
View Full Product Details