Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir33
- Entrez Gene ID
-
100314214
- Gene Name
- microRNA mir-33
- Gene Aliases
-
rno-mir-33
- Chromosome Location
-
Chr.7: 123431612 - 123431680 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_031847.1 |
|
1 - 1 |
35 |
68 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
coiled-coil domain containing 134 |
Ccdc134 |
|
-54.7kb
|
Sense |
Chr. - 123362469 - 123376930 on Build Rnor_6.0 |
Downstream |
shisa family member 8 |
Shisa8 |
RGD1563996 |
8.5kb
|
Anti-sense |
Chr. - 123440163 - 123445613 on Build Rnor_6.0 |
View Full Product Details