Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntn5
Entrez Gene ID
308587
Gene Name
netrin 5
Gene Aliases
RGD1565197
Chromosome Location
Chr.1: 101667693 - 101678112 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173526.1 NP_001166997.1 1 - 2 85 99
XM_008759380.1 XP_008757602.1 2 - 3 1206 99
XM_008759381.1 XP_008757603.1 3 - 4 1056 99

View Full Product Details