Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grxcr2
Entrez Gene ID
681048
Gene Name
glutaredoxin, cysteine rich 2
Gene Aliases
-
Chromosome Location
Chr.18: 36308925 - 36322320 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191078.1 NP_001178007.1 1 - 2 339 95

View Full Product Details