Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyb5d1
Entrez Gene ID
363629
Gene Name
cytochrome b5 domain containing 1
Gene Aliases
RGD1559567
Chromosome Location
Chr.10: 55993206 - 55997426 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191890.1 NP_001178819.1 3 - 4 451 99
XM_006246767.2 XP_006246829.1 4 - 5 503 99
GenBank mRNA FQ223142.1 - 3 - 4 842 99

View Full Product Details