Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcdc2c
Entrez Gene ID
500637
Gene Name
doublecortin domain containing 2C
Gene Aliases
RGD1559651
Chromosome Location
Chr.6: 47738716 - 47812224 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195807.1 NP_001182736.1 6 - 7 806 92
XM_006239955.2 XP_006240017.1 6 - 7 1177 92
XM_006239956.2 XP_006240018.1 6 - 7 1176 92
XM_006239957.2 XP_006240019.1 6 - 7 1176 92
XM_006239958.2 XP_006240020.1 6 - 7 1176 92

View Full Product Details